Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium trifluoromethanesulfonate

Chemical Identifiers

Chemical Name or Material Barium trifluoromethanesulfonate
CAS 2794-60-7
Formula Weight 435.48
MDL Number MFCD00143644
UN Number UN1564
Beilstein 3729346
Synonym Barium triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3857104

  SDS
alfa aesar™
3857104
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3857109

  SDS
alfa aesar™
3857109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3857118

  SDS
alfa aesar™
3857118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 435.48
Chemical Name or Material Barium trifluoromethanesulfonate
TSCA TSCA
Physical Form Powder
MDL Number MFCD00143644
Sensitivity Hygroscopic
CAS 2794-60-7
UN Number UN1564
Beilstein 3729346
Synonym Barium triflate
Health Hazard 1 Warning
Melting Point >300°C
Quantity 2g
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.