Please login to your online account to display your discounted pricing

Barium trifluoroacetate hydrate, White crystalline, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Barium trifluoroacetate hydrate
Physical Form White crystals
Molecular Formula Ba(OOCCF3)2·xH2O
Formula Weight 363.37g/mol
MDL Number MFCD00137143
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4459609

 
alfa aesar
44596-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4459618

 
alfa aesar
44596-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Anhydrous or Hydrate Hydrate
Color White
Quantity 10g
Chemical Name or Material Barium trifluoroacetate hydrate
Physical Form White crystals
Molecular Formula Ba(OOCCF3)2·xH2O
Formula Weight 363.37g/mol
MDL Number MFCD00137143
Health Hazard 1 H302-H332: Harmful if swallowed. Harmful if inhaled.
DOT Information DOT Class: 6.1, Packaging Group: III
Safety and Handling

H302-H332: Harmful if swallowed. Harmful if inhaled.