Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium titanium oxide sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis excluding Sr)

Chemical Identifiers

Chemical Name or Material Barium titanium oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Molecular Formula BaTiO3
MDL Number MFCD00003447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44806KS

 
alfa aesar™
44806KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Barium titanium oxide sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.9%
Assay Percent Notes (metals basis excluding Sr)
Molecular Formula BaTiO3
MDL Number MFCD00003447
Quantity 1 each
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.