missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium titanium oxide, Puratronic™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1065222

 View more versions of this product

Catalog No. AA1065222



Specifications

Specifications

Barium titanium oxide
(metals basis)
14,1000
233.19
99.99%
5.85 g/mL
White
100g
12047-27-7
MFCD00003447
Barium titanate
Powder
Puratronic™
Odorless
1620°C

Chemical Identifiers

12047-27-7
MFCD00003447
Barium titanate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 234-975-0

RTECSNumber : XR1437333

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents