missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium titanium oxide, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01065214

 View more versions of this product

Catalog No. AA1065214



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Barium titanium oxide
12047-27-7
Powder
(metals basis)
MFCD00003447
Barium titanate
[Ba++].[O-][Ti]([O-])=O
233.19
99.99%
Odorless
1620°C
White
25 g
BaO3Ti
14,1000
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
233.19
Puratronic™
5.85 g/mL

Chemical Identifiers

12047-27-7
233.19
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
BaO3Ti
MFCD00003447
Barium titanate
[Ba++].[O-][Ti]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 234-975-0

RTECSNumber : XR1437333

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only