Learn More
Barium titanium oxide, 99%, Thermo Scientific Chemicals
Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers
Supplier: Thermo Scientific Chemicals 01234818
Description
Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium titanium oxide | |
12047-27-7 | |
50 g | |
BaO3Ti | |
14,1000 | |
Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids | |
[Ba++].[O-][Ti]([O-])=O | |
233.19 | |
99% | |
5.85 g/mL |
1620°C | |
Powder | |
(metals basis) | |
MFCD00003447 | |
Barium titanate | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) | |
233.19 | |
Odorless |
Chemical Identifiers
12047-27-7 | |
233.19 | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) |
BaO3Ti | |
MFCD00003447 | |
Barium titanate | |
[Ba++].[O-][Ti]([O-])=O |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 234-975-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only