missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium titanium oxide, 99.9% (metals basis)

For vacuum deposition

Manufacturer:  Alfa Aesar™ 8826622

 View more versions of this product

Catalog No. AA8826622



Specifications

Specifications

Barium titanium oxide
99.9%
BaTiO3
14,1000
233.19
5.85g/mL
3 to 12mm
100g
12047-27-7
(metals basis)
MFCD00003447
Barium titanate
Pieces
Odorless
1620°C

Chemical Identifiers

12047-27-7
BaTiO3
MFCD00003447
Barium titanate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents