missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium titanium oxide, 99.7% (metals basis)

Manufacturer:  Alfa Aesar™ 8826736

 View more versions of this product

Catalog No. AA8826736



Specifications

Specifications

Barium titanium oxide
(metals basis)
MFCD00003447
Barium titanate
Powder
5.85g/mL
<2 Micron
1620°C
12047-27-7
BaTiO3
14,1000
233.19
99.7%
Odorless
White
500g

Chemical Identifiers

12047-27-7
BaTiO3
MFCD00003447
Barium titanate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XR1437333

TSCA : TSCA

SDS Documents