Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol

Chemical Identifiers

Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide
Name Note Produces 5% BaTiO3, in isopropanol
Molecular Formula C23H50BaO7Ti
Formula Weight 623.89
MDL Number MFCD01310249
UN Number UN1219
Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43893AC

  SDS
alfa aesar™
43893AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43893AP

  SDS
alfa aesar™
43893AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 623.89
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Physical Form Liquid
Name Note Produces 5% BaTiO3, in isopropanol
Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide
MDL Number MFCD01310249
Assay ≥99.5%
Molecular Formula C23H50BaO7Ti
UN Number UN1219
Health Hazard 1 Danger
Assay Percent Notes (metals basis)
Sensitivity Moisture Sensitive
Quantity 25mL
Health Hazard 2 GHS H Statement
H225-H361-H302-H332-H336
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.
Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide
Health Hazard 3 GHS P Statement
P210-P280h-P302+P352
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/clothing.
IF ON SKIN: Wash with plenty of soap and water.
Safety and Handling

Danger

GHS P Statement
P210-P280h-P302+P352
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/clothing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H225-H361-H302-H332-H336
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.