Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium thiosulfate, 98%

Chemical Identifiers

Chemical Name or Material Barium thiosulfate
CAS 35112-53-9
Molecular Formula BaO3S2
Linear Formula BaS2O3
Formula Weight 249.46
MDL Number MFCD00040661
UN Number UN1564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1498114

 
alfa aesar™
A1498114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1498122

 
alfa aesar™
A1498122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1498136

 
alfa aesar™
A1498136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.46
Chemical Name or Material Barium thiosulfate
TSCA TSCA
Assay 98%
Molecular Formula BaO3S2
Linear Formula BaS2O3
MDL Number MFCD00040661
CAS 35112-53-9
UN Number UN1564
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P302+P352
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P302+P352
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.