missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium thiosulfate, 98%

Manufacturer:  Alfa Aesar™ A1498122

 View more versions of this product

Catalog No. AAA1498122



Specifications

Specifications

Barium thiosulfate
BaO3S2
MFCD00040661
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
[O-]S(=O)(=S)[O-].[Ba+2]
249.444
249.46
100g
35112-53-9
BaS2O3
UN1564
ONPIOWQPHWNPOQ-UHFFFAOYSA-L
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
169663
98%

Chemical Identifiers

35112-53-9
249.444
ONPIOWQPHWNPOQ-UHFFFAOYSA-L
169663
[O-]S(=O)(=S)[O-].[Ba+2]
BaO3S2
MFCD00040661
barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P302+P352
IF ON SKIN: Wash with plenty of soap and water.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Documents