Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium sulfide, tech.

Chemical Identifiers

Chemical Name or Material Barium sulfide
CAS 21109-95-5
Purity Grade Technical Grade
Molecular Formula BaS
Formula Weight 169.40g/mol
MDL Number MFCD00014192
UN Number UN3134
Merck Index 14,995
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1342218

 
alfa aesar™
A13422-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Like Rotten Eggs
Melting Point 1200°C
Quantity 50g
Chemical Name or Material Barium sulfide
CAS 21109-95-5
Purity Grade Technical Grade
Molecular Formula BaS
Formula Weight 169.40g/mol
MDL Number MFCD00014192
UN Number UN3134
Merck Index 14,995
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H261-H302
Toxic if inhaled.
In contact with water releases flammable gas.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P231+P232-P261-P280-P304+P340-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
TSCA TSCA

Applications
Barium sulfide is used in brown patinas as well as for dehairing hides, in flame retardants, vulcanized rubber and luminous paints.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P231+P232-P261-P280-P304+P340-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H261-H302
Toxic if inhaled.
In contact with water releases flammable gas.
Harmful if swallowed.