missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium sulfate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1065106

 View more versions of this product

Catalog No. AA1065106



Specifications

Specifications

Barium sulfate
(metals basis)
BaSO4
14,994
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium(2+);sulfate
24414
Powder
Puratronic™
Odorless
1580°C (decomposition)
7727-43-7
BaO4S
MFCD00003455
barite
[O-]S(=O)(=O)[O-].[Ba+2]
233.383
233.4
99.998%
4.5g/mL
White
5g

Chemical Identifiers

7727-43-7
233.383
TZCXTZWJZNENPQ-UHFFFAOYSA-L
24414
[O-]S(=O)(=O)[O-].[Ba+2]
BaO4S
MFCD00003455
barite
barium(2+);sulfate
Safety and Handling

Safety and Handling

RTECSNumber : CR0600000

TSCA : TSCA

Documents