missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium rod, 22mm (0.9 in.) dia., 99+% (metals basis), Sr =0.8%

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

Manufacturer:  Alfa Aesar™ 1010322

 View more versions of this product

Catalog No. AA1010322



Description

Description

  • Packaged under argon
Specifications

Specifications

Barium rod
100g
7440-39-3
(metals basis)
MFCD00134031
and compounds
[Ba]
Odorless
5355457
Rod
Gray
≈7.3cm/100g
22mm (0.9 in.) dia.
Sr ≤0.8%
Ba
UN1400
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
barium
137.327
CHEBI:32595
≥99%

Chemical Identifiers

7440-39-3
137.327
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
5355457
barium
Ba
MFCD00134031
and compounds
CHEBI:32595
[Ba]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378i-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: BARIUM

RTECSNumber : CQ8370000

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents