Please login to your online account to display your discounted pricing

Barium, Reference Standard Solution (1000ppm ± 1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Ba) Solute: Barium Chloride Solvent: Distilled Water

Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSB162100

 View more versions of this product

Catalog No. SB162-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Quantity 100mL
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Chemical Name or Material Barium, Reference Standard Solution
Name Note 1000ppm ± 1%
Concentration 990 to 1010ppm
MDL Number MFCD00134031
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
CAS 7732-18-5
CAS 10361-37-2
ChemAlert Storage Symbol Gray
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED BARIUM REF STD

Specifications

Chemical Name or Material Barium, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 10361-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134031
Accessories
 

SDS & Safety Documents