Please login to your online account to display your discounted pricing

Barium, Reference Standard Solution (1000ppm ± 1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Ba) Solute: Barium Chloride Solvent: Distilled Water

$81.35 - $88.53

Chemical Identifiers

Chemical Name or Material Barium, Reference Standard Solution
Name Note 1000ppm ± 1%
CAS 10361-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134031
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB162-500

  SDS
fisher scientific company
SB162500
Poly Bottle 500mL Each for $81.35
Case of 6 Each for $488.10

SB162-100

  SDS
fisher scientific company
FLSB162100
Poly Bottle 100mL Each for $88.53
Case of 6 Each for $531.18
Description & Specifications

Specifications

Melting Point 0°C
Purity Grade Certified Grade
Quantity 500mL
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Color Colorless
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Concentration 990 to 1010ppm
ChemAlert Storage Symbol Gray
CAS 10361-37-2
MDL Number MFCD00134031
CAS 7732-18-5
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED BARIUM REF STD
Name Note 1000ppm ± 1%
Chemical Name or Material Barium, Reference Standard Solution
Safety and Handling