missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13876AE

 View more versions of this product

Catalog No. AA13876AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Barium
Clear liquid
MFCD00134031
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+);dinitrate
24798
23 hPa (17mm Hg) at 20°C
100mL
10022-31-8,7732-18-5,7732-18-5
BaN2O6
Nitric acid: 5.0%; Barium nitrate: 0.19%; Water: 94.81%
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
261.335
Liquid
Colorless

Chemical Identifiers

10022-31-8
261.335
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+);dinitrate
BaN2O6
MFCD00134031
24798
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents