missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium peroxide, White powder, 84%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1400818

 View more versions of this product

Catalog No. AA1400818



Specifications

Specifications

Barium peroxide
84%
MFCD00003454
ZJRXSAYFZMGQFP-UHFFFAOYSA-N
barium(2+);peroxide
14773
Powder
Odorless
Anhydrous
50g
1304-29-6
BaO2
barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
[O-][O-].[Ba+2]
169.325
169.34
4.96g/cm3
Moisture sensitive
White

Chemical Identifiers

1304-29-6
169.325
ZJRXSAYFZMGQFP-UHFFFAOYSA-N
14773
[O-][O-].[Ba+2]
BaO2
MFCD00003454
barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
barium(2+);peroxide
Safety and Handling

Safety and Handling

Keep away from food, drink and animal feeding stuffs. Avoid contact with skin and eyes. Take off immediately all contaminated clothing.

H272-H301-H331: May intensify fire; oxidizer. Toxic if swallowed. Toxic if inhaled.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 215-128-4

Documents