Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium perchlorate trihydrate, 99+%

Chemical Identifiers

Chemical Name or Material Barium perchlorate trihydrate
CAS 10294-39-0
Molecular Formula Ba(ClO4)2·3H2O
Formula Weight 390.29 (336.24 Anhydrous)
MDL Number MFCD00149148
UN Number UN1447
Merck Index 14,987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1759822

  SDS
alfa aesar™
A1759822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 390.29 (336.24 Anhydrous)
Health Hazard 3 GHS P Statement
P221-P283-P210-P306+P360-P371+P380+P375-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 10294-39-0
Merck Index 14,987
Molecular Formula Ba(ClO4)2·3H2O
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM PERCHLORATE, SOLID
Odor Odorless
Assay ≥99%
Quantity 100g
UN Number UN1447
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.
TSCA TSCA
Chemical Name or Material Barium perchlorate trihydrate
MDL Number MFCD00149148
Safety and Handling

Danger

GHS P Statement
P221-P283-P210-P306+P360-P371+P380+P375-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.