Please login to your online account to display your discounted pricing

Barium Perchlorate, Certified, 0.0100N ±0.0005N (0.005M), LabChem™

$51.03 - $51.03

Chemical Identifiers

Chemical Name or Material Barium Perchlorate
CAS 7732-18-5
CAS Max % 99.83
CAS 13465-95-7
CAS Max % 0.17
Purity Grade Certified Grade
Linear Formula Ba(ClO4)2
Formula Weight 336.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC116452

 
lab chem
LC116452
1L Each for $51.03
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 336.23
Concentration 0.0100N ±0.0005N (0.005M)
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Density 1g/mL
Linear Formula Ba(ClO4)2
CAS 7732-18-5
Identification Passes Test
Color Clear, colorless
CAS Max % 99.83
Specific Gravity 1g/mL
CAS 13465-95-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.17
Quantity 1L
Solubility Soluble in water
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Chemical Name or Material Barium Perchlorate
Physical Form Liquid
Decomposition Information Chlorine; Barium
Packaging Amber Glass
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.