Learn More
Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221275000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Barium perchlorate | |
99% | |
White | |
500 g | |
BaCl2O8 | |
MFCD00003440 | |
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv | |
OOULUYZFLXDWDQ-UHFFFAOYSA-L | |
barium(2+);diperchlorate | |
61623 | |
99% | |
Glass bottle | |
3.2000g/mL |
505.0°C | |
13465-95-7 | |
Powder | |
98.5% min. (Complexometry) | |
Ba(ClO4)2 | |
15, 983 | |
Solubility in water: soluble. Other solubilities: soluble in methanol, slightly soluble in ethanol, ethyl acetate and, acetone | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] | |
336.23 | |
336.23 | |
Extra Pure | |
3.2 |
Chemical Identifiers
13465-95-7 | |
336.23 | |
OOULUYZFLXDWDQ-UHFFFAOYSA-L | |
61623 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2] |
BaCl2O8 | |
MFCD00003440 | |
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv | |
barium(2+);diperchlorate |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep/Store away from clothing/other combustible materials.
Take any precaution to avoid mixing with combustibles.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 236-710-4
RTECSNumber : SC7550000
TSCA : TSCA
RUO – Research Use Only