Please login to your online account to display your discounted pricing

Barium Oxide, Powder (Tech.), 90%, ACROS Organics™

$86.43 - $86.43

Chemical Identifiers

Chemical Name or Material Barium oxide
CAS 1304-28-5
CAS 513-77-9
CAS 1304-29-6
Purity Grade Extra Pure
Assay ≥88% (HCl-titr.)
Assay Percent Range 90%
Molecular Formula BaO
Linear Formula BaO
Formula Weight 153.34
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC207835000

  SDS
thermo fisher scientific
207835000
Plastic bottle 500g Each for $86.43

AC207830250

  SDS
thermo fisher scientific
AC207830250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC207830025

  SDS
thermo fisher scientific
AC207830025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Extra Pure
Physical Form Powder
Assay ≥88% (HCl-titr.)
Formula Weight 153.34
Color Light yellow-greenish to grayish
Quantity 500g
Particle Size < 13 Micron
Iron (Fe) 170ppm
Chemical Name or Material Barium oxide
Packaging Plastic bottle
Aluminum (Al) 500ppm
Silicon Dioxide 0.67%
Sodium (Na) 320ppm
Molecular Formula BaO
Linear Formula BaO
Sulfur (S) 300ppm max.
Trace Analysis Trace analysis are typical values
Health Hazard 1 Danger
Barium Carbonate 11% max.
CAS 1304-29-6
Assay Percent Range 90%
CAS 1304-28-5
CAS 513-77-9
Safety and Handling

Danger