Learn More
Barium oxide, 90%, extra pure, powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207830025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2000.0°C | |
1920.0°C | |
Gray to Green-Yellow | |
2.5 kg | |
BaO | |
MFCD00003453 | |
15,982 | |
Solubility in water: soluble. Other solubilities: soluble in diluted acids, alcohols, insoluble in acetone, nh3, slowly but considerably soluble in methanol,, ethanol forming barium alcoholate | |
O=[Ba] | |
153.33 | |
153.34 | |
Extra Pure |
Barium oxide | |
1304-28-5,513-77-9,1304-29-6 | |
Powder | |
90% | |
BaO | |
01,683; 09,23 | |
barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum | |
QVQLCTNNEUAWMS-UHFFFAOYSA-N | |
oxobarium | |
62392 | |
90% | |
Plastic bottle |
Chemical Identifiers
1304-28-5 | |
153.33 | |
QVQLCTNNEUAWMS-UHFFFAOYSA-N | |
62392 | |
O=[Ba] |
BaO | |
MFCD00003453 | |
barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum | |
oxobarium |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 215-127-9
RUO – Research Use Only