missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium oxalate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1300906

 View more versions of this product

Catalog No. AA1300906



Specifications

Specifications

Barium oxalate
(metals basis)
BaC2O4
UN1564
barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
C(=O)(C(=O)[O-])[O-].[Ba+2]
225.345
225.36
99.999%
2.658g/mL
White
5g
516-02-9
C2BaO4
MFCD00054429
14,985
GXUARMXARIJAFV-UHFFFAOYSA-L
barium(2+);oxalate
68201
Powder
Puratronic™
Odorless
400°C (decomposition)

Chemical Identifiers

516-02-9
225.345
GXUARMXARIJAFV-UHFFFAOYSA-L
68201
C(=O)(C(=O)[O-])[O-].[Ba+2]
C2BaO4
MFCD00054429
barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
barium(2+);oxalate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Documents