missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Barium Nitrate, ACS Reagent, ≥99%, Honeywell™ Fluka™

ACS Reagent,≥99%

$120.80 - $120.80

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.335
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
IUPAC Name barium(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045272 View Documents Honeywell Fluka™
217581100G
100g Poly Bottle Each for $120.80
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10022-31-8
261.335
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
24798
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
BaN2O6
MFCD00003442
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
barium(2+);dinitrate

Specifications

10022-31-8
Ba(NO3)2
UN1446
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
261.335
261.34g/mol
ACS Reagent
Poly Bottle
592°C
Barium Nitrate
BaN2O6
MFCD00003442
Crystals
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+);dinitrate
24798
≥99%
3.24g/cu.cm
Colorless
100g
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P305 + P351 + P338-P370 + P378

H272-H302 + H332-H319

DOTInformation : 6.1(5.1)/PGII

EINECSNumber : 233-020-5

RTECSNumber : CQ9625000