Learn More
Barium nitrate, ACS, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012194A3
Description
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium nitrate | |
10022-31-8 | |
Crystalline | |
>99% | |
MFCD00003442 | |
Soluble in water. Insoluble in alcohol. | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
261.34 | |
261.35 | |
Hygroscopic | |
3.24 |
592°C (decomposition) | |
White | |
2 kg | |
BaN2O6 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
barium(2+);dinitrate | |
24798 | |
ACS | |
White crystalline |
Chemical Identifiers
10022-31-8 | |
261.34 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
24798 |
BaN2O6 | |
MFCD00003442 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H272-H301-H319-H332
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 233-020-5
RTECSNumber : CQ9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only