Learn More
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
$40.08 - $1099.36
Chemical Identifiers
CAS | 10022-31-8 |
---|---|
Molecular Formula | BaN2O6 |
Molecular Weight (g/mol) | 261.34 |
MDL Number | MFCD00003442 |
InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
Synonym | barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 |
PubChem CID | 24798 |
SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1064806
|
Thermo Scientific Chemicals
01064806 |
5 g |
Each for $40.08
|
|
|||||
AA1064814
|
Thermo Scientific Chemicals
01064814 |
25 g |
Each for $107.56
|
|
|||||
AA1064822
|
Thermo Scientific Chemicals
01064822 |
100 g |
Each for $294.82
|
|
|||||
AA1064836
|
Thermo Scientific Chemicals
01064836 |
500 g |
Each for $1,099.36
|
|
|||||
Description
Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener.Â
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10022-31-8 | |
261.34 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
24798 |
BaN2O6 | |
MFCD00003442 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
592°C (decomposition) | |
White | |
5 g | |
BaN2O6 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
barium(2+) dinitrate | |
24798 | |
Hygroscopic | |
3.24 g/cm3 |
10022-31-8 | |
Crystalline | |
99.99% (Metal basis) | |
MFCD00003442 | |
Soluble in water. Slightly soluble in ethanol and acetone. Partially soluble in cold water. | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
261.34 | |
261.35 | |
White crystalline | |
Barium nitrate |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H272-H301-H319-H332
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 233-020-5
RTECSNumber : CQ9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only