missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium nitrate, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3048122

 View more versions of this product

Catalog No. AA3048122



Specifications

Specifications

Barium nitrate
99.95%
MFCD00003442
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
261.335
261.35
White
10022-31-8
BaN2O6
White crystalline
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium(2+);dinitrate
24798
Crystalline
100g

Chemical Identifiers

10022-31-8
261.335
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
24798
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
BaN2O6
MFCD00003442
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
barium(2+);dinitrate
Safety and Handling

Safety and Handling

H272-H302-H332

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 233-020-5

SDS Documents