Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium molybdenum oxide, 99.9% (metals basis excluding Sr)

Electronic and optical equipment, pigment in paints and other protective coatings

Chemical Identifiers

Chemical Name or Material Barium molybdenum oxide
CAS 7787-37-3
Formula Weight 297.28
MDL Number MFCD00014182
UN Number UN1564
Synonym Barium molybdate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4020514

  SDS
alfa aesar™
4020514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4020522

  SDS
alfa aesar™
4020522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.28
Physical Form Powder
Assay Percent Notes (metals basis excluding Sr)
MDL Number MFCD00014182
Synonym Barium molybdate
Color White
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Chemical Name or Material Barium molybdenum oxide
TSCA TSCA
Mesh Size −100 Mesh
Odor Odorless
Assay 99.9%
CAS 7787-37-3
UN Number UN1564
Melting Point ≈1600°C
Quantity 25g
Solubility Information Slightly soluble in water and acids
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.98g/mL
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.