missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium metaphosphate, 90+%

Manufacturer:  Alfa Aesar™ 2264136

 View more versions of this product

Catalog No. AA2264136



Specifications

Specifications

Barium metaphosphate
BaO6P2
UN1564
Insoluble in water; Soluble in acids by a slow dissolution
[O-]P(=O)=O.[O-]P(=O)=O.[Ba+2]
16164194
Powder
Odorless
500g
13762-83-9
MFCD00061417
barium metaphosphate, barium 2+ ; dioxido oxo phosphanium, ba.2o3p, bismetaphosphoric acid barium salt, barium 2+ bis trioxidophosphate 1-
XNJIKBGDNBEQME-UHFFFAOYSA-L
295.269
295.25
≥90%
1560°C

Chemical Identifiers

13762-83-9
295.269
XNJIKBGDNBEQME-UHFFFAOYSA-L
16164194
BaO6P2
MFCD00061417
barium metaphosphate, barium 2+ ; dioxido oxo phosphanium, ba.2o3p, bismetaphosphoric acid barium salt, barium 2+ bis trioxidophosphate 1-
[O-]P(=O)=O.[O-]P(=O)=O.[Ba+2]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

SDS Documents