Please login to your online account to display your discounted pricing

Barium Metaborate Monohydrate, +90%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC381965000

 View more versions of this product

Catalog No. AC381965000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥90.0%
Formula Weight 240.97
Physical Form Powder
Quantity 500g
Color White
Purity Grade Pure
Packaging Plastic bottle
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Barium metaborate monohydrate
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Molecular Formula B2BaO4·H2O
Assay Percent Range 90+%
MDL Number MFCD00717540
CAS 19004-06-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Barium metaborate monohydrate
CAS 19004-06-9
Purity Grade Pure
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula B2BaO4·H2O
Formula Weight 240.97
MDL Number MFCD00717540
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents