Please login to your online account to display your discounted pricing

Barium manganate(VI), 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  198480250

 View more versions of this product

Catalog No. AC198480250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Fine Crystalline Powder
Formula Weight 256.27
Quantity 25g
Color Blue to dark blue-black
Merck Index 15, 977
EINECS Number 232-109-6
TSCA TSCA
Fieser 08,21; 09,23; 10,16; 12,38; 15,18; 16,18
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 95%
CAS 7787-35-1
Linear Formula BaMnO4
Chemical Name or Material Barium manganate(VI)
Molecular Formula BaMnO4
MDL Number MFCD00011514
Name Note 95%
Packaging Glass bottle

Specifications

Chemical Name or Material Barium manganate(VI)
Name Note 95%
CAS 7787-35-1
Assay ≥94%
Assay Percent Range 95%
Molecular Formula BaMnO4
Linear Formula BaMnO4
Formula Weight 256.27
MDL Number MFCD00011514
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.

SDS & Safety Documents