Learn More
Barium manganate(VI), 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 198480050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Barium manganate(VI) | |
7787-35-1 | |
94% min | |
BaMnO4 | |
08,21; 09,23; 10,16; 12,38; 15,18; 16,18 | |
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese | |
ZZCNKSMCIZCVDR-UHFFFAOYSA-N | |
barium(2+);dioxido(dioxo)manganese | |
3084030 | |
95% |
95% | |
5g | |
BaMnO4 | |
MFCD00011514 | |
15, 977 | |
Solubility in water: slightly soluble. Other solubilities: soluble in acids | |
[Ba++].[O-][Mn]([O-])(=O)=O | |
256.26 | |
256.27 | |
Glass bottle |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 232-109-6
TSCA : TSCA
RUO â Research Use Only