missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium manganate(VI), 95%, ACROS Organics™

$144.93 - $317.35

Chemical Identifiers

CAS 7787-35-1
Molecular Formula BaMnO4
Molecular Weight (g/mol) 256.27
MDL Number MFCD00011514
InChI Key ZZCNKSMCIZCVDR-UHFFFAOYSA-N
Synonym acmc-1bdpw
PubChem CID 3084030
IUPAC Name barium(2+);dioxido(dioxo)manganese
SMILES [O-][Mn](=O)(=O)[O-].[Ba+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC198480050 SDS View Documents Thermo Fisher Scientific
AC198480050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC198480250 SDS View Documents Thermo Fisher Scientific
198480250
25g Glass bottle Each for $144.93
 
AC198481000 SDS View Documents Thermo Fisher Scientific
198481000
100g Glass bottle Each for $317.35
 
Specifications

Chemical Identifiers

7787-35-1
256.27
ZZCNKSMCIZCVDR-UHFFFAOYSA-N
3084030
[O-][Mn](=O)(=O)[O-].[Ba+2]
BaMnO4
MFCD00011514
acmc-1bdpw
barium(2+);dioxido(dioxo)manganese

Specifications

7787-35-1
BaMnO4
MFCD00011514
15, 977
Solubility in water: slightly soluble. Other solubilities: soluble in acids
[O-][Mn](=O)(=O)[O-].[Ba+2]
256.27
256.27
95%
Black-Blue to Blue
Barium manganate(VI), 95%
94% min
BaMnO4
08,21; 09,23; 10,16; 12,38; 15,18; 16,18
acmc-1bdpw
ZZCNKSMCIZCVDR-UHFFFAOYSA-N
barium(2+);dioxido(dioxo)manganese
3084030
Fine Crystalline Powder
Glass bottle
5g
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 232-109-6

TSCA : TSCA