Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium isopropoxide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium isopropoxide
CAS 24363-37-9
Linear Formula Ba[OCH(CH3)2]2
Formula Weight 255.52
MDL Number MFCD00050486
UN Number UN3205
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4127114

  SDS
alfa aesar™
4127114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4127103

  SDS
alfa aesar™
4127103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4127106

  SDS
alfa aesar™
4127106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Barium isopropoxide
CAS 24363-37-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
Physical Form Powder/Lump
Assay 99.5%
Assay Percent Notes (metals basis)
Linear Formula Ba[OCH(CH3)2]2
Formula Weight 255.52
MDL Number MFCD00050486
Sensitivity Moisture Sensitive
UN Number UN3205
Color White
Melting Point ≈200°C (dec.)
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.