Learn More
Barium isopropoxide, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01452314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Barium isopropoxide | |
Brown to Yellow | |
Powder | |
C6H14BaO2 | |
MFCD00050486 | |
barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
CC(C)[O-].CC(C)[O-].[Ba+2] | |
255.503 | |
255.52 |
∼200°C (decomposition) | |
24363-37-9 | |
25 g | |
Ba[OCH(CH3)2]2 | |
UN3205 | |
CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
barium(2+);propan-2-olate | |
6101105 | |
Moisture sensitive |
Chemical Identifiers
24363-37-9 | |
255.503 | |
CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
6101105 | |
CC(C)[O-].CC(C)[O-].[Ba+2] |
C6H14BaO2 | |
MFCD00050486 | |
barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
barium(2+);propan-2-olate |
Safety and Handling
GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.
P210-P235+P410-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H252-H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only