missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium iodide, White beads, 99.995% (Metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Chemical Identifiers

CAS 13718-50-8
Physical Form Beads
Assay Percent Range 99.995% (Metals basis)
Molecular Formula BaI2
Formula Weight 391.15
MDL Number MFCD00003452
Chemical Name or Material Barium iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3567906 SDS Alfa Aesar
3567906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3567914 SDS Alfa Aesar
3567914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13718-50-8
99.995% (Metals basis)
391.15
Barium iodide
Beads
BaI2
MFCD00003452

Specifications

Beads
5g
BaI2
Odorless
99.995% (Metals basis)
391.15
White
MFCD00003452
13718-50-8
Barium iodide

Safety and Handling

H302-H332: Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 237-276-9