Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium iodide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Barium iodide
CAS 13718-50-8
Name Note Ultra dry
Formula Weight 391.15
MDL Number MFCD00003452
UN Number UN1564
Merck Index 14,980
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4711814

  SDS
alfa aesar™
4711814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 391.15
Packaging Ampule
Chemical Name or Material Barium iodide
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Assay 99.999%
Sensitivity Light Sensitive & Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00003452
CAS 13718-50-8
UN Number UN1564
Merck Index 14,980
Color White
Melting Point 740°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 5.15g/mL
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

  • Ampouled under argon