Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium iodate monohydrate

Chemical Identifiers

Chemical Name or Material Barium iodate monohydrate
CAS 7787-34-0
Linear Formula Ba(IO3)2·H2O
Formula Weight 505.15 (487.11 Anhydrous)
MDL Number MFCD00239354
UN Number UN3087
Merck Index 14,979
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3912918

  SDS
alfa aesar™
3912918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 505.15 (487.11 Anhydrous)
Health Hazard 3 GHS P Statement
P221-P210-P220-P261-P304+P340-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.998g/mL
TSCA TSCA
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
Physical Form Powder
MDL Number MFCD00239354
CAS 7787-34-0
Chemical Name or Material Barium iodate monohydrate
Odor Weak
Quantity 50g
Merck Index 14,979
Health Hazard 2 GHS H Statement
H272-H302-H332
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.
UN Number UN3087
Linear Formula Ba(IO3)2·H2O
Health Hazard 1 Danger
Solubility Information Insoluble in alcohol; Soluble in water, nitric acid, hydrochloric acid