Please login to your online account to display your discounted pricing

Barium Hydroxide, Saturated Solution, LabChem™

$190.31 - $190.31

Chemical Identifiers

Chemical Name or Material Barium Hydroxide
CAS 7732-18-5
CAS Max % 96.85
CAS 12230-71-6
CAS Max % 3.15
Name Note Saturated Solution
Linear Formula Ba(OH)2·8H2O
Formula Weight 315.47
UN Number UN3266
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC116402

 
lab chem
LC116402
1L Each for $190.31
Description & Specifications

Specifications

Formula Weight 315.47
CAS 7732-18-5
Color Clear, colorless
Name Note Saturated Solution
CAS Max % 3.15
Packaging Poly Bottle
CAS 12230-71-6
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Quantity 1L
Chemical Name or Material Barium Hydroxide
Storage Information Room Temperature
UN Number UN3266
Decomposition Information Barium
Physical Form Liquid
Health Hazard 1 Danger
Identification Passes Test
Solubility Soluble in water
Specific Gravity 1.03g/mL
CAS Max % 96.85
Density 1.03g/mL
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Linear Formula Ba(OH)2·8H2O
Safety and Handling

Danger

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.