Please login to your online account to display your discounted pricing

Barium Hydroxide, Saturated, Fisher Chemical


Quantity 250mL
Packaging Poly Bottle

Manufacturer: ricca chemical company  SB9100250

 View more versions of this product

Catalog No. SB9100-250

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 250mL
DOT Information DOT Class 8, : Corrosive
Methods For AOAC Method 937.05
CAS Min % 91.0
CAS Max % 94.0
Packaging Poly Bottle
ChemAlert Storage Symbol White
CAS 12230-71-6
CAS Max % 7.56
CAS Min % 7.26
Chemical Name or Material Barium Hydroxide, Saturated
CAS 7732-18-5

Specifications

Chemical Name or Material Barium Hydroxide, Saturated
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 94.0
CAS 12230-71-6
CAS Min % 7.26
CAS Max % 7.56
Accessories