Please login to your online account to display your discounted pricing

Barium Hydroxide Octahydrate (Crystalline/Certified ACS), Fisher Chemical

$321.31 - $321.31

Chemical Identifiers

Chemical Name or Material Barium Hydroxide Octahydrate
CAS 12230-71-6
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥98.0%
Assay Percent Range ≥98.0 %
Molecular Formula H2BaO2 · 8 H2O
Linear Formula Ba(OH)2·8H2O
Formula Weight 315.46
MDL Number MFCD00149152
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B46-500

  SDS
fisher scientific company
FLB46500
Glass Bottle 500g Each for $321.31
Case of 6 Each for $1,927.86
Description & Specifications

Specifications

Assay ≥98.0%
Melting Point 78°C
Purity Grade Certified ACS Grade
Quantity 500g
Health Hazard 3 Emergency Overview

Causes burns by all exposure routes. Ingestion causes severe swelling, severe damage to the delicate tissue and danger of perforation: Product is a corrosive material. Use of gastric lavage or emesis is contraindicated. Possible perforation of stomach or esophagus should be investigated. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
Health Hazard 2 Danger
Formula Weight 315.46
UN Number 2923
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula H2BaO2 · 8 H2O
pH 12.5
Color White
Density 2.180g/cm³
Calcium (Ca) 0.05% max.
Chloride 0.001% max.
Heavy Metals (as Pb) 5ppm max.
Assay Percent Range ≥98.0 %
Sodium (Na) 0.01% max.
Potassium (K) 0.01% max.
Chemical Name or Material Barium Hydroxide Octahydrate
Linear Formula Ba(OH)2·8H2O
CAS 12230-71-6
MDL Number MFCD00149152
Identification Pass Test
Packaging Glass Bottle
Iron (Fe) 0.001% max.
Carbonate 2.0% max.
Sulfide Pass Test
Strontium (Sr) 0.8% max.
Insoluble in Hydrochloric Acid 0.01% max.
ChemAlert Storage Symbol White

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview

Causes burns by all exposure routes. Ingestion causes severe swelling, severe damage to the delicate tissue and danger of perforation: Product is a corrosive material. Use of gastric lavage or emesis is contraindicated. Possible perforation of stomach or esophagus should be investigated. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1

Danger