Please login to your online account to display your discounted pricing

Barium hydroxide octahydrate, 98+%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC203140050

 View more versions of this product

Catalog No. AC203140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystals, Crystalline Flakes or Powder
Formula Weight 315.48
Quantity 5kg
Color White
Purity Grade For analysis
Density 2.1800g/mL
Merck Index 15, 973
Iron (Fe) 5ppm max.
Packaging Plastic bottle
Assay Percent Range 98+%
Molecular Formula H2BaO2·8H2O
Linear Formula Ba(OH)2·8H2O
CAS 12230-71-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Barium hydroxide octahydrate
Heavy Metals (as Pb) 5ppm max.
Insoluble Matter 0.01% max. (in HCl)
Sulfur (S) 5ppm max.
MDL Number MFCD00149152
Trace Analysis 1.5% max.
Health Hazard 1 GHS Signal Word: Danger
Barium Carbonate 2% max.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Calcium (Ca) 50ppm max.
Chlorine (Cl) 20ppm max.

Specifications

Chemical Name or Material Barium hydroxide octahydrate
CAS 12230-71-6
Purity Grade For analysis
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula H2BaO2·8H2O
Linear Formula Ba(OH)2·8H2O
Formula Weight 315.48
MDL Number MFCD00149152
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.