missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Barium Hydroxide Monohydrate, purum p.a., ≥98.0% (T), Honeywell™

Purum p.a.,≥98.0% (T)

$127.73 - $234.82

Chemical Identifiers

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide monohydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044706 View Documents Honeywell™
11783100G
100g Poly Bottle Each for $127.73
 
60044707 View Documents Honeywell™
117831KG
1kg Poly Bottle Each for $234.82
 
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

22326-55-2
189.356
GKQTUHKAQKWLIN-UHFFFAOYSA-L
16211219
O.[OH-].[OH-].[Ba+2]
BaH4O3
MFCD00149151
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide monohydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
barium(2+);dihydroxide;hydrate

Specifications

22326-55-2
Ba(OH)2 · H2O
UN3262
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide monohydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
O.[OH-].[OH-].[Ba+2]
189.356
189.36g/mol
purum p.a.
Poly Bottle
300 hPa at 78°C
White
100g
BaH4O3
MFCD00149151
Powder
GKQTUHKAQKWLIN-UHFFFAOYSA-L
barium(2+);dihydroxide;hydrate
16211219
≥98.0% (T)
3.743g/mL at 25°C
40g/L at 20°C
780°C
13.1 (21.0g/L at 25°C)
Barium Hydroxide Monohydrate
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H302 + H332-H314

DOTInformation : 8/PGIII

EINECSNumber : 241-234-5