missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Barium hydroxide monohydrate, 95%

Manufacturer:  Alfa Aesar™ 4588822

 View more versions of this product

Catalog No. AA4588822



Specifications

Specifications

Barium hydroxide monohydrate
BaH4O3
MFCD00149151
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide monohydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
O.[OH-].[OH-].[Ba+2]
189.356
189.38 (171.36 Anhydrous)
95%
Odorless
White
22326-55-2
Ba(OH)2·H2O
UN3262
GKQTUHKAQKWLIN-UHFFFAOYSA-L
barium(2+);dihydroxide;hydrate
16211219
Crystalline
3.743g/mL
Air Sensitive & Hygroscopic
100g

Chemical Identifiers

22326-55-2
189.356
GKQTUHKAQKWLIN-UHFFFAOYSA-L
16211219
O.[OH-].[OH-].[Ba+2]
BaH4O3
MFCD00149151
barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide monohydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
barium(2+);dihydroxide;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.

RTECSNumber : CQ9200000

SDS Documents