Learn More
Barium hydroxide octahydrate, 99.95%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 377820050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Barium hydroxide octahydrate | |
12230-71-6 | |
Powder, Crystals and/or Chunks | |
99.95% | |
Ba(OH)2·8H2O | |
15, 973 | |
Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] | |
315.46 | |
315.48 | |
Trace Metal Basis | |
2.18 |
78.0°C | |
White | |
5 g | |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
barium(2+);dihydroxide;octahydrate | |
17749109 | |
99.95% | |
Glass bottle | |
2.1800g/mL |
Chemical Identifiers
12230-71-6 | |
315.46 | |
ZUDYPQRUOYEARG-UHFFFAOYSA-L | |
17749109 | |
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++] |
BaH18O10 | |
MFCD00149152 | |
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate | |
barium(2+);dihydroxide;octahydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only