Please login to your online account to display your discounted pricing

Barium Hydroxide Anhydrous (Powder/Certified), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLB47500

 View more versions of this product

Catalog No. B47-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Purity Grade Certified Grade
Formula Weight 171.34
Melting Point 78°C
Quantity 500g
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula BaH2O2
Viscosity [NULL]
Color Clear
Density 2.18g/cm³
Packaging Glass Bottle
Chemical Name or Material Barium Hydroxide Anhydrous
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Insoluble in Hydrochloric Acid 0.01% max.
Iron (Fe) 0.005% max.
Assay Percent Range ≥95.0 %
Linear Formula Ba(OH)2
Sulfide 0.001% max.
MDL Number MFCD00003443
CAS 17194-00-2
ChemAlert Storage Symbol White
Calcium and Alkalis 1.3% max.
Chloride 0.01% max.

Specifications

Chemical Name or Material Barium Hydroxide Anhydrous
CAS 17194-00-2
Purity Grade Certified Grade
Physical Form Solid
Assay ≥95.0%
Assay Percent Range ≥95.0 %
Molecular Formula BaH2O2
Linear Formula Ba(OH)2
Formula Weight 171.34
MDL Number MFCD00003443
Accessories