Please login to your online account to display your discounted pricing

Barium Hydroxide Anhydrous (Powder/Certified), Fisher Chemical

$384.85 - $400.92

Chemical Identifiers

Chemical Name or Material Barium Hydroxide Anhydrous
CAS 17194-00-2
Purity Grade Certified Grade
Physical Form Solid
Assay ≥95.0%
Assay Percent Range ≥95.0 %
Molecular Formula BaH2O2
Linear Formula Ba(OH)2
Formula Weight 171.34
MDL Number MFCD00003443
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B47-250

  SDS
fisher scientific company
FLB47250
Glass Bottle 250g Each for $400.92
Case of 6 Each for $2,405.52

B47-500

  SDS
fisher scientific company
FLB47500
Glass Bottle 500g Each for $384.85
Case of 6 Each for $2,309.10
Description & Specifications

Specifications

Assay ≥95.0%
Purity Grade Certified Grade
Formula Weight 171.34
Melting Point 78°C
Quantity 250g
DOT Information DOT Class 8, : Corrosive
Physical Form Solid
Molecular Formula BaH2O2
Color Clear
Density 2.18g/cm³
Chemical Name or Material Barium Hydroxide Anhydrous
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Insoluble in Hydrochloric Acid 0.01% max.
Iron (Fe) 0.005% max.
Packaging Glass Bottle
Assay Percent Range ≥95.0 %
Linear Formula Ba(OH)2
Sulfide 0.001% max.
MDL Number MFCD00003443
CAS 17194-00-2
ChemAlert Storage Symbol White
Calcium and Alkalis 1.3% max.
Chloride 0.01% max.
Safety and Handling