Please login to your online account to display your discounted pricing

Barium Hydroxide, Anhydrous, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  612425000

 View more versions of this product

Catalog No. AC612425000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Formula Weight 141.35
Quantity 500g
EINECS Number 241-234-5
TSCA TSCA
Density 10.9
RTECS Number CQ9200000
Merck Index 15, 973
Chloride 0.01% max.
Additional Information Vapor Pressure: Negligible
Calcium and Alkalis 1.30% max.
Chemical Name or Material Barium hydroxide
Heavy Metals (as Pb) 0.001% max.
Iron (Fe) 0.005% max.
Molecular Formula BaH2O2
Linear Formula Ba(OH)2
MDL Number MFCD00003443
CAS 17194-00-2
Identification Pass Test
Insoluble in Hydrochloric Acid 0.010% max.
Sulfide 0.001% max.

Specifications

Chemical Name or Material Barium hydroxide
Assay ≥95.0%
Molecular Formula BaH2O2
Linear Formula Ba(OH)2
Formula Weight 141.35
MDL Number MFCD00003443