Please login to your online account to display your discounted pricing

Barium Hydroxide, Anhydrous, ACROS Organics™


Quantity 250g

Manufacturer: thermo fisher scientific  612422500

 View more versions of this product

Catalog No. AC612422500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Formula Weight 141.35
Quantity 250g
Merck Index 15, 973
TSCA TSCA
EINECS Number 241-234-5
RTECS Number CQ9200000
Density 10.9
Heavy Metals (as Pb) 0.001% max.
Iron (Fe) 0.005% max.
Insoluble in Hydrochloric Acid 0.010% max.
Identification Pass Test
Linear Formula Ba(OH)2
Molecular Formula BaH2O2
MDL Number MFCD00003443
Chemical Name or Material Barium hydroxide
CAS 17194-00-2
Chloride 0.01% max.
Additional Information Vapor Pressure: Negligible
Sulfide 0.001% max.
Calcium and Alkalis 1.30% max.

Specifications

Chemical Name or Material Barium hydroxide
Assay ≥95.0%
Molecular Formula BaH2O2
Linear Formula Ba(OH)2
Formula Weight 141.35
MDL Number MFCD00003443